Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: syg-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129687.6 | NP_001123159.1 | 8 - 9 | 1077 | 115 | |
NM_001373246.4 | NP_001360786.1 | 9 | 1078 | 115 | ||
NM_076056.8 | NP_508457.3 | 8 - 9 | 1071 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |