Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vang-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076099.5 | NP_508500.1 | 4 - 5 | 652 | 94 | |
NM_076100.7 | NP_508501.1 | 4 - 5 | 629 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |