Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twk-26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076121.7 | NP_508522.2 | 8 - 9 | 1402 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |