Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sec-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076129.6 | NP_508530.1 | 1 - 2 | 143 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |