Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lbp-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047785.4 | NP_001041250.1 | 3 - 4 | 384 | 72 | |
NM_001383542.1 | 11 - 12 | 3090 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Inventoried Probe spans exons |