Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dhs-27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076190.5 | NP_508591.3 | 4 - 5 | 569 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |