Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rgs-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029543.5 | NP_001024714.1 | 9 - 10 | 1754 | 66 | |
NM_001029544.6 | NP_001024715.1 | 5 - 6 | 645 | 66 | ||
NM_001373260.4 | NP_001360776.1 | 11 | 1928 | 66 | ||
NM_001373261.4 | NP_001360777.1 | 10 - 11 | 1919 | 66 | ||
NM_001373262.3 | NP_001360778.1 | 10 | 1839 | 66 | ||
NM_001374994.1 | NP_001361946.1 | 2 - 3 | 345 | 66 | ||
NM_076209.7 | NP_508610.3 | 10 - 11 | 1919 | 66 | ||
GenBank mRNA | AB080201.1 | - | 9 - 10 | 1785 | 66 | |
AY569308.1 | - | 10 - 11 | 1830 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |