Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: abu-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076258.6 | NP_508659.1 | 5 - 6 | 783 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |