Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fkh-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029589.4 | NP_001024760.1 | 2 - 3 | 332 | 95 | |
NM_001029590.6 | NP_001024761.1 | 1 - 2 | 238 | 95 | ||
NM_001380939.1 | NP_001367812.1 | 3 - 4 | 453 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Caenorhabditis elegans Probe spans exons |