Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ttr-48 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076265.4 | NP_508666.1 | 3 - 4 | 267 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Caenorhabditis elegans Probe spans exons |