Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sup-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076273.5 | NP_508674.1 | 4 - 5 | 486 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |