Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lin-18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076283.6 | NP_508684.4 | 4 - 5 | 482 | 91 | |
GenBank mRNA | AF133217.1 | - | 4 - 5 | 537 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Caenorhabditis elegans Probe spans exons |