Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acr-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076291.5 | NP_508692.3 | 10 - 11 | 1449 | 148 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |