Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clh-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029714.2 | NP_001024885.1 | 6 - 7 | 1291 | 121 | |
NM_001029715.3 | NP_001024886.1 | 6 - 7 | 1254 | 121 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |