Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clh-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029714.2 | NP_001024885.1 | 15 - 16 | 2497 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |