Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vha-12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076310.8 | NP_508711.1 | 1 - 2 | 237 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |