Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twk-28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076331.6 | NP_508732.3 | 11 - 12 | 1522 | 94 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |