Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dop-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029407.4 | NP_001024578.1 | 3 - 4 | 443 | 78 | |
NM_001029408.5 | NP_001024579.1 | 2 - 3 | 354 | 78 | ||
NM_001383561.1 | 3 - 4 | 1053 | 78 |
Set Membership: |
Amplicon length between 71 and 85 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |