Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029194.4 | NP_001024365.1 | 10 - 11 | 1455 | 73 | |
NM_001380949.1 | NP_001367445.1 | 11 - 12 | 1380 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Recommended Assay Caenorhabditis elegans Probe spans exons |