Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-35 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029194.4 | NP_001024365.1 | 6 - 7 | 938 | 103 | |
NM_001380949.1 | NP_001367445.1 | 7 - 8 | 863 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |