Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ham-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029192.6 | NP_001024363.2 | 5 - 6 | 937 | 66 | |
NM_001029193.7 | NP_001024364.2 | 5 - 6 | 942 | 66 | ||
NM_001307898.3 | NP_001294827.1 | 4 - 5 | 750 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |