Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lnp-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076387.6 | NP_508788.1 | 5 - 6 | 821 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Inventoried Probe spans exons |