Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: spat-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029733.6 | NP_001024904.2 | 14 - 15 | 5385 | 63 | |
NM_001029734.4 | NP_001024905.1 | 14 - 15 | 5387 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |