Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: aman-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076410.4 | NP_508811.1 | 12 - 13 | 1552 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |