Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tyra-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029633.5 | NP_001024804.1 | 7 - 8 | 1164 | 88 | |
NM_001029634.3 | NP_001024805.1 | 7 - 8 | 1303 | 88 | ||
NM_001047810.5 | NP_001041275.1 | 5 - 6 | 857 | 88 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |