Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lfi-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001375027.2 | NP_001362033.1 | 10 - 11 | 4491 | 102 | |
NM_001375029.2 | NP_001362032.1 | 9 - 10 | 4284 | 102 | ||
NM_001375030.2 | NP_001362152.1 | 9 - 10 | 4442 | 102 | ||
NM_001375031.2 | NP_001362153.1 | 8 - 9 | 4192 | 102 | ||
NM_001383570.1 | 10 - 11 | 4795 | 102 | |||
NM_076447.6 | NP_508848.2 | 11 - 12 | 4829 | 102 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |