Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ets-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001277027.4 | NP_001263956.1 | 6 - 7 | 675 | 86 | |
NM_001383573.1 | 6 - 7 | 712 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |