Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vps-41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380962.1 | NP_001367568.1 | 2 - 3 | 405 | 130 | |
NM_001383575.1 | 2 - 3 | 410 | 130 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |