Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: jkk-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383579.1 | 8 - 9 | 1086 | 79 | ||
GenBank mRNA | AB024086.1 | - | 8 - 9 | 1026 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |