Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sulp-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076543.6 | NP_508944.1 | 10 - 11 | 1405 | 75 | |
GenBank mRNA | AY887908.1 | - | 10 - 11 | 1427 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |