Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rgs-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307891.3 | NP_001294820.1 | 10 - 11 | 1451 | 93 | |
NM_001307892.4 | NP_001294821.1 | 8 - 9 | 1608 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Caenorhabditis elegans Probe spans exons |