Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hbl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001047778.5 | NP_001041243.1 | 4 - 5 | 2046 | 75 | |
NM_001047779.5 | NP_001041244.1 | 3 - 4 | 1845 | 75 | ||
GenBank mRNA | AF097737.1 | - | 4 - 5 | 2037 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |