Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mig-23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076593.8 | NP_508994.1 | 7 - 8 | 1354 | 83 | |
GenBank mRNA | AB126261.1 | - | 7 - 8 | 1334 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |