Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kin-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029671.6 | NP_001024842.1 | 3 - 4 | 426 | 85 | |
NM_001083302.4 | NP_001076771.1 | 4 - 5 | 530 | 85 | ||
NM_001380985.1 | NP_001367669.1 | 4 - 5 | 514 | 85 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |