Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hsp-25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029203.3 | NP_001024374.1 | 5 - 6 | 731 | 56 | |
NM_001380987.1 | NP_001367469.1 | 5 - 6 | 614 | 56 | ||
NM_001383593.1 | 5 - 6 | 2602 | 56 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |