Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hsp-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307849.4 | NP_001294778.1 | 1 - 2 | 395 | 66 | |
NM_001383595.1 | 4 - 5 | 1496 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Inventoried Probe spans exons |