Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mom-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076653.5 | NP_509054.1 | 5 - 6 | 888 | 74 | |
GenBank mRNA | AF013489.1 | - | 5 - 6 | 885 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |