Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: igcm-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001375051.2 | NP_001362087.1 | 10 - 11 | 1284 | 115 | |
NM_001375052.3 | NP_001362088.1 | 9 - 10 | 1231 | 115 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |