Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nas-33 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076685.4 | NP_509086.2 | 14 - 15 | 1484 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |