Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ets-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001373331.1 | NP_001359862.1 | 3 - 4 | 385 | 92 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |