Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dop-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029736.6 | NP_001024907.2 | 1 - 2 | 415 | 119 | |
NM_001029737.5 | NP_001024908.2 | 1 - 2 | 213 | 119 | ||
NM_001038472.5 | NP_001033561.1 | 1 - 2 | 301 | 119 | ||
NM_001381012.1 | NP_001368078.1 | 1 - 2 | 181 | 119 | ||
GenBank mRNA | AY485309.1 | - | 1 - 2 | 209 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Caenorhabditis elegans Probe spans exons |