Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acr-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076727.9 | NP_509128.1 | 10 - 11 | 1528 | 133 | |
GenBank mRNA | X86403.1 | - | 1407 | 133 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |