Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acr-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076728.4 | NP_509129.1 | 9 - 10 | 1057 | 66 | |
GenBank mRNA | Y08637.1 | - | 1057 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |