Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tyra-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029350.7 | NP_001024521.1 | 4 - 5 | 517 | 103 | |
NM_001029351.6 | NP_001024522.1 | 3 - 4 | 281 | 103 | ||
NM_001038448.5 | NP_001033537.1 | 3 - 4 | 289 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |