Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hen-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076799.6 | NP_509200.1 | 4 - 5 | 309 | 77 | |
GenBank mRNA | AB067680.1 | - | 4 - 5 | 331 | 77 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |