Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mek-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029600.6 | NP_001024771.1 | 4 - 5 | 907 | 80 | |
NR_132545.1 | 3 - 4 | 562 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |