Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ceh-14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076872.4 | NP_509273.1 | 6 - 7 | 587 | 66 | |
GenBank mRNA | AF244368.1 | - | 6 - 7 | 601 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |