Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029435.5 | NP_001024606.1 | 4 - 5 | 454 | 139 | |
NR_136373.1 | 3 - 4 | 468 | 139 | |||
GenBank mRNA | S66176.1 | - | 4 - 5 | 418 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |