Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kvs-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001029318.5 | NP_001024489.1 | 10 - 11 | 1359 | 84 | |
NM_001029319.4 | NP_001024490.1 | 10 - 11 | 1333 | 84 | ||
NM_001373368.2 | NP_001360715.1 | 11 - 12 | 1390 | 84 | ||
NM_001375071.1 | NP_001361979.1 | 10 - 11 | 1318 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |