Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mec-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_076912.5 | NP_509313.1 | 3 - 4 | 960 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |